missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Square Narrow-Mouth PPCO Bottles with Closure

Catalog Number 10054870
31.35 EUR valid until 2024-12-31
Use promo code "21983" to get your promotional price.


Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
500 mL
1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Easily measure and dispense using these translucent, square, narrow-mouth PPCO bottles, featuring mouths wider than conventional narrow-mouth bottles.

Easily measure and dispense using Thermo Scientific™ Nalgene™ Square Narrow-Mouth PPCO Bottles with Closure, featuring mouths wider than conventional narrow-mouth bottles. These polypropylene copolymer bottles are graduated, repeatedly autoclavable, translucent, and resistant to alcohols and bases. They are made only from high-quality, laboratory-grade, plastic materials for dependably low leachables and extractables.

Features:
  • Square sides allow for good use of space and an easy grip
  • Graduated for ease of dispensing
  • Linerless caps provide the ultimate in leakproof† protection without the use of a liner that can wrinkle, cause leaks or contaminate your reagents
  • Autoclavable/Leakproof†
  • Translucent bottles provide visible liquid level
  • Useful between 0° and 135°C (32° and 275°F)

Includes:

  • Leakproof† polypropylene screw caps

Note:

Remove cap or completely disengage threads before autoclaving. Bottle neck design is protected by US Trademark Reg. No. 2857283. Square bottle with arched shoulders design is protected by US Trademark Reg. No. 2857279.

TRUSTED_SUSTAINABILITY
Capacity 4 oz., 125 mL
Closure Material Polypropylene
Closure Size 38-430 mm
Closure Type Screw Cap
Diameter (Metric) 53 mm
Height (English) 4.3 in.
Height (Metric) 109 mm
Through the Closure Dispensing No
Width (English) 2.12 in.
Product Type Narrow Mouth Bottle
Capacity (Metric) 125 mL
Capacity (English) 4 oz.
Color Translucent
Description Narrow-Mouth Square Bottle
Diameter (English) 2.1 in.
Diameter (English) Inner Neck 1.1 in.
Diameter (Metric) Inner Neck 28 mm
Includes Leakproof polypropylene screw closures
Material PPCO
No. per Case 72
No. per Pack 12
Sterility Non-sterile
Closure Color Natural, translucent
Graduations Yes
Certifications/Compliance USP Class VI; FDA - 21 CFR
Closure Included Yes
Autoclavable Autoclavable
Graduated Yes
Mouth Narrow
Shape Square
Show More Show Less

† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'