Learn More
Thermo Scientific™ Nalgene™ Tubing T-Type Connectors
Maximize liquid flow with non-tapered hose barbed T-Type Connectors, molded of polypropylene resin, free of animal derived components.
20.82 € - 185.00 €
Specifications
Product Type | Tubing Connector |
---|---|
Autoclavable | Autoclavable |
Material | Polypropylene |
Product Code | Brand | For Use With | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With | Price | Quantity & Availability | |||||
15231169
|
Thermo Scientific™
6151-0125 |
Tubing I.D. 1/8 in. |
156.00 €
Pack of 72 |
In Stock
Log in to see stock. |
|||||
15243069
|
Thermo Scientific™
6151-0187 |
Tubing I.D. 3/16 in. |
141.00 €
Pack of 72 |
In Stock
Log in to see stock. |
|||||
15253069
|
Thermo Scientific™
6151-0250 |
Tubing I.D. 1/4 in. |
185.00 €
Pack of 72 |
In Stock
Log in to see stock. |
|||||
10258690
|
Thermo Scientific™
6151-0250 |
Tubing I.D. 1/4 in. |
25.70 €
Pack of 12 |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
15263069
|
Thermo Scientific™
6151-0312 |
Tubing I.D. 5/16 in. |
175.00 €
Pack of 72 |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
10320681
|
Thermo Scientific™
6151-0312 |
Tubing I.D. 5/16 in. |
29.70 €
Pack of 12 |
In Stock
Log in to see stock. |
|||||
10015050
|
Thermo Scientific™
6151-0375 |
Tubing I.D. 3/8 in. |
44.80 €
Pack of 12 |
In Stock
Log in to see stock. |
|||||
15283069
|
Thermo Scientific™
6151-0500 |
Tubing I.D. 1/2 in. |
171.00 €
Pack of 48 |
In Stock
Log in to see stock. |
|||||
15273069
|
Thermo Scientific™
6151-0375 |
Tubing I.D. 3/8 in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10106700
|
Thermo Scientific™
6151-0125 |
Tubing I.D. 1/8 in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10310681
|
Thermo Scientific™
6151-0187 |
Tubing I.D. 3/16 in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Maximize liquid flow with non-tapered hose barbed Thermo Scientific™ Nalgene™ Tubing T-Type Connectors, molded of polypropylene resin, free of animal derived components. Products are double-bagged to maintain cleanliness.
- Autoclavable
Specifications
Tubing Connector | |
Autoclavable | |
Polypropylene |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.