Learn More
Thermo Scientific™ Nalgene™ Tubing Y-Type Connectors
Maximize liquid flow with non-tapered hose barbed Y-Type Connectors, molded of polypropylene resin, free of animal derived components.
21.85 € - 217.00 €
Specifications
Product Type | Tubing Connector |
---|---|
Autoclavable | Autoclavable |
Material | Polypropylene |
Product Code | Brand | For Use With | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With | Price | Quantity & Availability | |||||
10684402
|
Thermo Scientific™
6152-0125 |
1/8 in. I.D. |
33.80 €
Pack of 12 |
In Stock
Log in to see stock. |
|||||
11704743
|
Thermo Scientific™
6152-0187 |
3/16 in. I.D. |
151.00 €
Pack of 72 |
In Stock
Log in to see stock. |
|||||
10186070
|
Thermo Scientific™
6152-0250 |
1/4 in. I.D. |
26.17 €
Pack of 12 |
In Stock
Log in to see stock. |
|||||
11850872
|
Thermo Scientific™
6152-0250 |
1/4 in. I.D. |
157.00 €
Pack of 72 |
In Stock
Log in to see stock. |
|||||
15203079
|
Thermo Scientific™
6152-0312 |
5/16 in. I.D. |
188.00 €
Pack of 72 |
In Stock
Log in to see stock. |
|||||
10020601
|
Thermo Scientific™
6152-0312 |
5/16 in. I.D. |
31.33 €
Pack of 12 |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
10470913
|
Thermo Scientific™
6152-0375 |
3/8 in. |
38.00 €
Pack of 12 |
In Stock
Log in to see stock. |
|||||
10778691
|
Thermo Scientific™
6152-0500 |
1/2 in. Tubing |
44.00 €
Pack of 12 |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
15213079
|
Thermo Scientific™
6152-0500 |
1/2 in. Tubing |
217.00 €
Pack of 48 |
In Stock
Log in to see stock. |
|||||
10471001
|
Thermo Scientific™
6152-0187 |
3/16 in. I.D. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15293069
|
Thermo Scientific™
6152-0125 |
1/8 in. I.D. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11384535
|
Thermo Scientific™
6152-0375 |
3/8 in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Maximize liquid flow with non-tapered hose barbed Thermo Scientific™ Nalgene™ Tubing Y-Type Connectors, molded of polypropylene resin, free of animal derived components. Products are double-bagged to maintain cleanliness.
- Polypropylene construction
- Autoclavable
Specifications
Tubing Connector | |
Autoclavable | |
Polypropylene |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.