Learn More
Thermo Scientific™ Nalgene™ Utility Boxes
Use these attractive, easy-to-clean boxes, available in four different sizes and shapes, for general-purpose laboratory storage.
24.68 €
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10792191
|
Thermo Scientific™
5700-2000 |
2000 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Use these attractive, easy-to-clean Thermo Scientific™ Nalgene™ Utility Boxes for general-purpose laboratory storage. Ideal for multi-purpose lab storage, boxes are available in four different sizes and shapes. Lids snap securely in place, clear walls offer easy view of contents and rounded corners allow for easy cleaning.
- Manufactured from clear styrene-acrylonitrile with tight-fitting, molded, white, LDPE lids that include inserts to store and keep items dry and secure
- Attractive and easy to clean
- Stackable for high-density storage
- Cat. No. 5700-0500 is ideal for electrophoresis gels
- Not leakproof
Specifications
Lid |