missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nandrolone Phenpropionate, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Nandrolone Phenpropionate |
| CAS | 62-90-8 |
| Analyte or Component Names | Nandrolone Phenpropionate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C27 H34 O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Nandrolone Phenpropionate, 19-Norandrostenolone phenylpropionate, 19-Nortestosterone phenylpropionate, Activin (anabolic steroid), Durabol, Durabolin, Fenobolin, NPP, NSC 23162, NTPP, Nandrobolic, Nandrolin, Nandrolon phenylpropionate, Nandrolone phenpropionate, Nandrolone phenylpropionate, Nerobil, Nerobolil, Norandrolone phenylpropionate, Norandrostenolone phenylpropionate, Nortestosterone phenylpropionate, Phenobolin, Strabolene, Superanabolon, Estr-4-en-3-one, 17-(1-oxo-3-phenylpropoxy)-, (17beta)-, Estr-4-en-3-one, 17beta-hydroxy-, hydrocinnamate (7CI,8CI), Estr-4-en-3-one, 17beta-hydroxy-, phenylpropionate (6CI), Hydrocinnamic acid, ester with 17beta-hydroxyestr-4-en-3-one (8CI), 17beta-(3-Phenylpropionyloxy)-estr-4-en-3-one, 17beta-Hydroxy-19-norandrost-4-en-3-one 17-phenylpropionate, 17beta-Hydroxy-4-estren-3-one17-phenylpropionate, 17beta-Hydroxyestr-4-en-3-one phenylpropionate, 19-Nor-17beta-hydroxy-3-ketoandrost-4-ene 17-phenylpropionate |
| InChI Formula | InChI=1S/C27H34O3/c1-27-16-15-22-21-11-9-20(28)17-19(21)8-10-23(22)24(27)12-13-25(27)30-26(29)14-7-18-5-3-2-4-6-18/h2-6,17,21-25H,7-16H2,1H3/t21-,22+,23+,24-,25-,27-/m0/s1 |
| Solution Type | Neat |
| SMILES | C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@H]34)[C@@H]1CC[C@@H]2OC(=O)CCc5ccccc5 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction