missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Nano-SIL ALUGRAM™ aluminum Sheets
Unmodified HPTLC silica layers on aluminum for TLC. Macherey-Nagel™ Nano-SIL ALUGRAM™ aluminum Sheets feature nano-silica 60 on state-of-the-art aluminum sheets that demonstrate low weight, high strength and temperature stability, and easy cutting.
263.00 €
Specifications
Particle Size | 2 to 10 μm |
---|---|
Pore Size | 60 Å |
Type | TLC Plate |
For Use With (Application) | HPTLC |
Pore Volume | 0.75 mL/g |
Product Code | Brand | Fluorescent Indicator | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Fluorescent Indicator | Price | Quantity & Availability | |||||
10185041
|
Macherey-Nagel™
818143 |
UV254 |
263.00 €
Pack of 25 |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
10093061
|
Macherey-Nagel™
818141 |
None |
263.00 €
Pack of 25 |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
Description
Unmodified HPTLC silica layers on aluminum for TLC. Macherey-Nagel™ Nano-SIL ALUGRAM™ aluminum Sheets feature nano-silica 60 on state-of-the-art aluminum sheets that demonstrate low weight, high strength and temperature stability, and easy cutting.
Unmodified HPTLC silica layers with High Definition properties.- Binder: highly polymeric product, which is stable in almost all organic solvents and resistant towards aggressive visualization reagents
- Narrow fractionation of the silica particles allows sharper separations, shorter developing times, shorter migration distances, smaller samples and an increased detection sensitivity compared to SIL G plates
- Fluorescent Indicator available: manganese activated zinc silicate with green fluorescence for short-wave UV (254 nm)
Specifications
2 to 10 μm | |
TLC Plate | |
0.75 mL/g | |
0.2 mm |
60 Å | |
HPTLC | |
∼500 m2/g | |
20 x 20 cm |