missing translation for 'onlineSavingsMsg'
Learn More

Naproxen sodium, 98%, Thermo Scientific™

Naproxen sodium (CAS # 26159-34-2) is an arylacetic acid of the non-steroidal anti-inflammatory drug class.

90.90 € - 814.00 €

Chemical Identifiers

CAS 26159-34-2
Molecular Formula C14H13NaO3
Molecular Weight (g/mol) 252.25
MDL Number MFCD00058507
InChI Key CDBRNDSHEYLDJV-FVGYRXGTSA-M
Synonym naproxen sodium, anaprox, miranax, naprelan, naproxen sodium salt, anapran, anaprotab, aprowell, floginex, flogogin
PubChem CID 23681059
ChEBI CHEBI:7477
IUPAC Name sodium (2S)-2-(6-methoxynaphthalen-2-yl)propanoate
SMILES [Na+].COC1=CC=C2C=C(C=CC2=C1)[C@H](C)C([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15475369
SDS View Documents
Thermo Scientific Alfa Aesar
J63103.06
5g
90.90 €
5g
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
15485369
SDS View Documents
Thermo Scientific Alfa Aesar
J63103.14
25g
276.00 €
25g
View alternative products
Save Up to 
»
Estimated Shipment: 04-09-2026
Add to basket
 
 Restricted Item
15495369
SDS View Documents
Thermo Scientific Alfa Aesar
J63103.22
100g
814.00 €
100g
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

General Description

  • Naproxen sodium is the sodium salt of naproxen
  • Naproxen sodium competitively suppresses prostaglandin formation and pyrogen release by blocking the conversion of arachidonic acid

Application

  • Naproxen is frequently used for migraine and postoperative pain research
  • It is suitable for cell culture and in vivo research on pain, inflammation, and fever
Specifications

Chemical Identifiers

26159-34-2
252.25
CDBRNDSHEYLDJV-FVGYRXGTSA-M
23681059
sodium (2S)-2-(6-methoxynaphthalen-2-yl)propanoate
C14H13NaO3
MFCD00058507
naproxen sodium, anaprox, miranax, naprelan, naproxen sodium salt, anapran, anaprotab, aprowell, floginex, flogogin
CHEBI:7477
[Na+].COC1=CC=C2C=C(C=CC2=C1)[C@H](C)C([O-])=O

Specifications

26159-34-2
MFCD00058507
naproxen sodium, anaprox, miranax, naprelan, naproxen sodium salt, anapran, anaprotab, aprowell, floginex, flogogin
CDBRNDSHEYLDJV-FVGYRXGTSA-M
sodium (2S)-2-(6-methoxynaphthalen-2-yl)propanoate
23681059
252.24
Naproxen sodium
C14H13NaO3
14,6417
Soluble in water up to 100mg/mlSoluble in methanol and chloroform; Slightly soluble in ether; Insoluble in water.
[Na+].COC1=CC=C2C=C(C=CC2=C1)[C@H](C)C([O-])=O
252.25
CHEBI:7477
98%
Videos
Safety and Handling

Safety and Handling

P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c

H302-H360

missing translation for 'einecsNumber' : 247-486-2

missing translation for 'rtecsNumber' : QJ1047000

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

Product Identifier
  • Naproxen sodium

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1A

Hazard Statement
  • H302-Harmful if swallowed.
  • H360-May damage fertility or the unborn child.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contain: Naproxen sodium

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.