missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ National Economy Vials

Thermo Scientific™ National Economy Vials are offered as clear or amber, in a range of styles to meet specific application needs.

Brand:  Thermo Scientific™ 60180-711

Code : A0

Additional Details : Weight : 3.00000kg

 View more versions of this product

Product Code. 13287489

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Thermo Scientific™ National Economy Vials are offered as clear or amber, in a range of styles to meet specific application needs.

Specifications

Specifications

60180-711
2 mL
Snap cap
Thermo Scientific, Agilent, PE, Shimadzu, Varian
Amber
2 mL
11 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers