Learn More
Thermo Scientific™ National™ Replacement Battery for Handheld Electronic Crimpers and Decappers
Rapid, efficient crimping and decapping of vials.
Brand: Thermo Scientific™ 60180-555
Code : NEW
Additional Details : Weight : 0.05000kg Transport : UN number : 3480 Chem class : 9
Description
Thermo Scientific™ National Handheld Electronic Crimpers and Decappers provide rapid, efficient crimping and decapping of vials.
- 7.2 V DC Replacement battery for Thermo Scientific National Handheld Electronic Crimper and Decapper.
Specifications
Replacement battery, 7.2V | |
National |