Learn More
Thermo Scientific™ National Unassembled Certified™ Vial Kits
Thermo Scientific™ National Unassembled Certified Vial Kits provide extra assurance that time after time, the product upholds rigorous manufacturing standards.
33.55 € - 116.55 €
Product Code | Brand | Volume (Metric) | Closure Material | Closure Type | Color | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Closure Material | Closure Type | Color | Price | Quantity & Availability | ||
11921543
|
Thermo Scientific™
CERT4000-71LV |
350 μL | Bonded PTFE, Silicone | Screw Cap | Clear |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
11981503
|
Thermo Scientific™
CERT4000-80 |
2 mL | PTFE, Rubber | Screw Cap | Clear |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
11981543
|
Thermo Scientific™
CERT400092W |
2 mL | PTFE, Silicone | Screw Cap | Clear |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
11951553
|
Thermo Scientific™
CERT4015-86 |
4 mL | Bonded PTFE, Silicone | Screw Cap | Amber |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
11961553
|
Thermo Scientific™
CERT4015-87 |
4 mL | Bonded PTFE, Silicone | Screw Cap | Amber |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
10514315
|
Thermo Scientific™
CERT4015-99 |
1 mL | Polyethylene | SepCap | Amber |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
Thermo Scientific™ National Unassembled Certified Vial Kits provide extra assurance that time after time, the product upholds rigorous manufacturing standards.
- Fully lot tested for HPLC and GC analysis
- Certificate of Conformance in every pack
- Bonded caps to prevent septa push-through
- Clear Type 1, 33 expansion borosilicate glass or amber Type 2, 51 expansion borosilicate glass
- Available with or without ID patch
- Fixed micro vials or inserts for limited-volume sampling