Learn More
NCOA2 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 30962-1-AP-20UL
 View more versions of this product
Product Code. 18034745
  • 145.00 € / 20µL
Description
The NCOA2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human NCOA2. This antibody recognizes human antigen. The NCOA2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
NCOA2
300 μg/mL
Rabbit
Primary
Human
150 kDa
IgG
Western Blot, ELISA
bHLHe75, GRIP1, hTIF2, KAT13C, NCoA 2, NCOA2, nuclear receptor coactivator 2, SRC2, TIF2
20 μL
10499
Store at -20°C. Stable for one year after shipment.