missing translation for 'onlineSavingsMsg'
Learn More

Nelociguat, MedChemExpress

Product Code. 16401989
Encompass_Preferred
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 16401989

Brand: Medchem Express HY78237

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Nelociguat (BAY60-4552) is a nitric oxide sensitive soluble guanylate cyclase stimulator.

Nelociguat (BAY60-4552) is a nitric oxide sensitive soluble guanylate cyclase stimulator.

Specifications

Chemical Name or Material Nelociguat
CAS 625115-52-8
Color Brown
Purity Grade Notes Research
Molecular Formula C19H17FN8O2
Quantity 1 g
Synonym BAY60-4552
Solubility Information DMSO : < 1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble or slightly soluble)
SMILES O=C(OC)NC1=C(N)N=C(C2=NN(CC3=CC=CC=C3F)C4=NC=CC=C42)N=C1N
Molecular Weight (g/mol) 408.39
Formula Weight 408.39
Percent Purity 99.13%
Grade Research
Physical Form Liquid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.