missing translation for 'onlineSavingsMsg'
Learn More

Neomycin sulfate hydrate, Thermo Scientific Chemicals

Catalog Number 15464099
Click to view available options
Quantity:
100 g
1405-10-3
C23H48N6O17S
712.72
MFCD29905465
OIXVKQDWLFHVGR-DJUBFIBWNA-N
124080932
OS(O)(=O)=O.NCC1OC(O[C@@H]2[C@@H](CO)O[C@@H](OC3C(O)C(N)CC(N)C3OC3OC(CN)C(O)C(O)C3N)[C@@H]2O)C(N)C(O)C1O
1405-10-3
C23H48N6O17S
712.72
MFCD29905465
OIXVKQDWLFHVGR-DJUBFIBWNA-N
124080932
OS(O)(=O)=O.NCC1OC(O[C@@H]2[C@@H](CO)O[C@@H](OC3C(O)C(N)CC(N)C3OC3OC(CN)C(O)C(O)C3N)[C@@H]2O)C(N)C(O)C1O

Neomycin sulfate is an aminoglycoside antibiotic and calcium channel protein inhibitor. Neomycin sulfate also binds to prokaryotic ribosomes inhibiting translation and is effective against gram-positive and gram-negative bacteria. Neomycin sulfate inhibits Phospholipase C via binding to inositol phospholipids. It also inhibits phosphatidylcholine-PLD activity and induces Ca2+ mobilization and PLA2 activation in human platelets. Neomycin sulfate inhibits DNase I induced DNA degradation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Neomycin sulfate is an aminoglycoside antibiotic and calcium channel protein inhibitor. Neomycin sulfate also binds to prokaryotic ribosomes inhibiting translation and is effective against gram-positive and gram-negative bacteria. Neomycin sulfate inhibits Phospholipase C via binding to inositol phospholipids. It also inhibits phosphatidylcholine-PLD activity and induces Ca2+ mobilization and PLA2 activation in human platelets. Neomycin sulfate inhibits DNase I induced DNA degradation.

Solubility
Soluble in water

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
TRUSTED_SUSTAINABILITY
Type Neomycin Sulfate
Physical Form Powder
Merck Index 14,6454
Solubility Information Soluble in water
IUPAC Name (2S,3R,4S,5S,6R)-5-amino-2-(aminomethyl)-6-[(1R,2R,3S,4R,6R)-4,6-diamino-2-[(2R,3S,4R,5S)-4-[(2S,3R,4S,5R,6S)-3-amino-6-(aminomethyl)-4,5-dihydroxyoxan-2-yl]oxy-3-hydroxy-5-(hydroxymethyl)oxolan-2-yl]oxy-3-hydroxycyclohexyl]oxyoxane-3,4-diol;sulfuric acid
Formula Weight 908.87 (Anhydrous)
Color White
Quantity 100 g
Chemical Name or Material Neomycin sulfate hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'