Learn More
Kartell™ Neoprene O-rings
Complete a filtration and vacuum desiccator with a spare o-ring. Kartell™ Neoprene O-rings form a seal with the transparent cover and polypropylene base of a filtration and vacuum desiccator.
Brand: Kartell™ 0055700
Code : A0
Additional Details : Weight : 70.41000kg
Description
- Essectial component of a Filtration and Vacuum Desiccator
- Spare O-ring
- Spread a little grease on O-ring for assistance in establishing initial vacuum
Specifications
O-Ring | |
1Each | |
For 8890A |
Black | |
Neoprene |