Learn More
Invitrogen™ Mouse LXR alpha (aa 35-49) Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP272
Code : LE
Additional Details : CAS Number : 69-74-9 Weight : 0.01000kg
Description
Mouse LXR alpha (residues 35-49) Synthetic Peptide, PEP-272, from Invitrogen.
PEP-272 is a 15 amino acid synthetic peptide whose sequence corresponds to residues 35-49 of mouse LXR alpha. The sequence of this peptide is (amino to carboxy terminus): T35-C-I-L-R-E-E-A-R-M-P-Q-S-T- G49This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product, LXR beta 2, catalog # PA1-332. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.Specifications
22259 | |
purified | |
Mouse LXR alpha (aa 35-49) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Mouse | |
Q9Z0Y9 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
Nr1h3 | |
No preservative | |
50 μg | |
T35 - C - I - L - R - E - E - A - R - M - P - Q - S - T- G49 |