Learn More
Thermo Scientific™ NF-kB Activation Inhibitor IV
Brand: Thermo Scientific™ J65706.MA
Code : NEW
Additional Details : Weight : 0.00010kg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- A cell-permeable trans-stilbene Resveratrol analog that exhibits much enhanced anti-inflammatory potency, but no anti-oxidant activity
Specifications
10 mg | |
2°C to 8°C | |
C15H13FO | |
(E)-2-Fluoro-4′-methoxystilbene | |
Soluble in DMSO or ethanol | |
Solid |
White | |
A cell-permeable trans-stilbene Resveratrol analog that exhibits much enhanced anti-inflammatory potency, but no anti-oxidant activity | |
228.26g/mol | |
Light Sensitive | |
Signal Transduction Reagent |