missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) octanoate, in mineral spirits (8% Ni), Thermo Scientific Chemicals

Catalog Number p-7050796
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
4995-91-9
C8H16NiO2
202.907
MFCD00064239
ANYMYNDNCKURBB-UHFFFAOYSA-N
nickel dioctanoate, nickel 2+ caprylate, nickel octoate, caprylic acid, nickel ii salt, octanoic acid, nickel 2+ salt
54611835
nickel;octanoic acid
CCCCCCCC(=O)O.[Ni]
This item is not returnable. View return policy
4995-91-9
C8H16NiO2
202.907
MFCD00064239
ANYMYNDNCKURBB-UHFFFAOYSA-N
nickel dioctanoate, nickel 2+ caprylate, nickel octoate, caprylic acid, nickel ii salt, octanoic acid, nickel 2+ salt
54611835
nickel;octanoic acid
CCCCCCCC(=O)O.[Ni]

Nickel(II) octanoate is used to prepare nickel based catalysts using niobia modified silica as support. It is also used as a catalyst and involved in the treatment of qaiyarah heavy crude oil, petrolene using ethanol as a solvent and reductant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Green
Density 0.95 g/mL
Flash Point 40°C (104°F)
Linear Formula [CH3(CH2)6COO]2Ni
Quantity 25 g
UN Number UN1993
Solubility Information Miscible with ethanol,chloroform,acetonitrile,ether,carbon disulfide,petroleum ether and glacial acetic acid. Immiscible with water.
Formula Weight 345.1
Concentration or Composition (by Analyte or Components) 8% Ni
Physical Form Liquid
Chemical Name or Material Nickel(II) octanoate, In mineral spirits
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'