Learn More
R&L Enterprises™ Hoffman Clips
An excellent option for sealing flexible tubing in the laboratory. R&L Enterprises™ Hoffman Clips are made of nickel plated brass to prevent corrosion.
Brand:  R&L Enterprises™ 9054
Code : 03
Additional Details : Weight : 0.50400kg
 View more versions of this product
Product Code. 12764396
  • 93.35 € / Pack of 10
Estimated Shipment: 19-06-2024
to see stock.
Add to basket
Description
Description
  • Either side or bottom opening
  • Available for a range of tube thicknesses
  • Threaded bar with thumbscrew
Specifications
Specifications
Clip
Nickel Plated
For use with 40mm O.D. tubing
10 Pack