missing translation for 'onlineSavingsMsg'
Learn More

Nickel Standard, 1000mg/L in 2% HNO3, (Assurance Grade), SPEX CertiPrep™

Catalog Number p-219084
Click to view available options
Quantity:
30 mL
250 mL
Packaging:
Poly Bottle
Triple-leached LDPE bottle
This item is not returnable. View return policy
7440-02-0, 7697-37-2, 7732-18-5
Ni
58.69
MFCD00011137 MFCD06798735
PXHVJJICTQNCMI-UHFFFAOYSA-N
935
CHEBI:28112
nickel
[Ni]
This item is not returnable. View return policy
7440-02-0, 7697-37-2, 7732-18-5
Ni
58.69
MFCD00011137 MFCD06798735
PXHVJJICTQNCMI-UHFFFAOYSA-N
935
CHEBI:28112
nickel
[Ni]

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ni1000μg/mL

Certifications

ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015

Compliance

GHS/OSHA Complaint

CAS Min % 0.1, 2.0, 97.9
CAS Max % 0.1, 2.0, 97.9
Grade Assurance
Concentration or Composition (by Analyte or Components) Matrix    Concentration    Cat. No.
2-5% HNO3    1000μg/mL    PLNI2-2Y, -2T, -2X    10,000    PLNI2-3Y, -3X
Packaging Triple-leached LDPE bottle
Solvent or Matrix 2% HNO3
Quantity 30 mL
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Chemical Name or Material Nickel Standard
compliance-icons compliance-icons
Product Identifier
  • 1000µg/ml nickel
Signal Word
  • Danger||Warning
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : nitric acid, nickel
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'