missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitric Acid 67-70%, Optima™, for Ultra Trace Elemental Analysis, Fisher Chemical™
615.65 EUR valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
250 mL
500 mL
1 L
Packaging:
FEP Bottle
Description
Nitric Acid, HNO3, CAS Number-7697-37-2, acide nitrique, acidum nitricum, azotic acid, rfna, aqua fortis, nitryl hydroxide, salpetersaeure, nitrous fumes, nital, hydrogen nitrate, 500mL
- Ideal for analysis by ICP-MS and high-resolution ICP-MS.
- Evaluated for 65 metals at low-level ppt concentrations.
- Manufactured, purified, and packaged in a Class 1000 clean room.
- Packaged in specially manufactured and cleaned FEP fluoropolymer bottles to maintain product integrity and to eliminate metal leaching from glass bottles.
- Three year shelf life from time of manufacture - available on COA.
Specifications
pH | 1 |
Color | Colorless Yellow |
Melting Point | -41°C |
Boiling Point | 120.5°C |
Density | 1.40 g/cm3 |
Packaging | FEP Bottle |
Vapor Pressure | 0.94kPa (20°C) |
Certificate of Analysis | Certificate of Lot Analysis is included with each bottle. |
Quantity | 1 L |
Physical Form | Liquid |
Show More |
Safety and Handling
Product Identifier
- Nitric acid 67-69%, Optima for Ultra traces analysis
- Danger
- Oxidising liquid Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- H272-May intensify fire; oxidiser.
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Nitric acid
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'