missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ BioClean™ Biotac™ Nitrile Gloves

Non-sterile cleanroom gloves with superior grip

Brand:  Ansell™ BIOTAC-L

Code : NEW

Additional Details : Weight : 8.80000kg

 View more versions of this product

Product Code. 15268168

  • 259.00 € / Pack of 1000

Estimated Shipment
06-07-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • 12 in.(300mm) Ambidextrous
  • AQL 1.5
  • Finger-textured, Powder-Free
  • Nitrile is resistant to a wide range of chemicals
  • Excellent ESD properties
  • 1.5 Freedom from Pinholes
  • Processed to ensure ISO Class 5 Compatibility
  • Latex-Free to eliminate Type-1 Latex Allergies
  • Category 3 - Complex Design Personal Protective Equipment
  • Complies with the requirements of the Glove Standard EN 420:2003, the European Medical Gloves Standard EN 455 (Parts 1, 2 & 3) & EN 374-1, 2 & 3:2003
Specifications

Specifications

Safety
Textured
Ambidextrous
100 pieces per sealed inner PE bag; one inner PE bag per sealed outer PE bag; 10 outer bags per lined carton (1000 pieces).
Powder-free
Non-sterile
ISO 9001:2015 and (EU)2016/425, EN420:2003 + A1:2009, EN374-1:20016, EN374-5:2016 and EN421:2010, EN455-2:2015, (EC)1935/2004 and (EC)2023/2006
ISO 5
Beaded/Rolled
Nitrile
Large
White
30.48 cm
Videos
Documents