missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitrofurantoin, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
25 g
100 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Yellow |
Infrared Spectrum | Authentic |
Assay Percent Range | 98% |
Quantity | 10 g |
IUPAC Name | 1-[(E)-(5-nitrofuran-2-yl)methylideneamino]imidazolidine-2,4-dione |
Formula Weight | 238.16 |
Percent Purity | ≥97.5% |
Physical Form | Powder |
Chemical Name or Material | Nitrofurantoin |
Safety and Handling
Product Identifier
- Nitrofurantoin
- Danger
- Acute toxicity Category 4
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'