missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitromethane, 96%, for HPLC, Thermo Scientific Chemicals
148.67 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 L
2.5 L
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -29.0°C |
Density | 1.1200g/mL |
Boiling Point | 100.0°C to 102.0°C |
Flash Point | 35°C |
Assay Percent Range | 95% min. (GC) |
Packaging | Glass bottle |
Linear Formula | CH3NO2 |
Refractive Index | 1.3805 to 1.3825 |
Quantity | 2.5 L |
Merck Index | 15,6698 |
Show More |
Safety and Handling
Product Identifier
- Nitromethane
- Warning
- Acute toxicity Category 4
- Flammable liquid Category 3
- H302-Harmful if swallowed.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'