missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitrosonium hexafluorophosphate, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Nitrosonium hexafluorophosphate is used as initiator in the polymerization reactions of 1,2-epoxides such as styrene oxide, cyclohexene oxide and 1,2-epoxybutane. Further, it is involved in the polymerization of styrene, n-butyl vinyl ether and tetrahydrofuran.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Crystalline |
Quantity | 5 g |
Linear Formula | NOPF6 |
UN Number | UN3260 |
Solubility Information | Soluble in acetonitrile. |
Formula Weight | 174.96 |
Percent Purity | 96% |
Odor | Odorless |
Sensitivity | Moisture sensitive |
Chemical Name or Material | Nitrosonium hexafluorophosphate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'