missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uvex™ NK4025B NBR-Coated Chemical Protection Gloves
Click to view available options
Size:
9
10
Description
- Multi-layered material structure ensures cut protection and resistance against chemical/mechanical hazards
- Layers are constructed from cotton, HPPE, and glass fiber
- NBR coating is resistant against grease, mineral oils, and a wide variety of chemicals
- Enhanced grip capabilities in damp, wet, and oily areas
- Cotton interlock lining for increased comfort
Specifications
Coating Material | NBR |
Color | Blue |
Material | HPPE, Polyamide |
Size | 9 |
EN388 Standard | 4X44C |
EN ISO 374-1 | Type A |
EN ISO 374-1 CHEMICAL LETTER CODES | JKNOPT |
For Use With (Application) | Handling wet, oily materials |
Glove Cuff Style | Gauntlet |
Liner Material | Interlock Cotton |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'