missing translation for 'onlineSavingsMsg'
Learn More

NMP, N-Methylpyrrolidone, GC Headspace Grade, Thermo Scientific™

Catalog Number 15552413
389.97 EUR valid until 2024-04-30
Use promo code "21741" to get your promotional price.


Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle

Processed to high purity for accurate and repeatable determination of trace levels of residual solvents

  • Every lot is tested to ensure interference free baselines for interference free determination of Class 1, 2, and 3 residual solvents
  • 1 ppm max. of Class 1 Solvents, 10 ppm max. of Class 2 solvents, and 50 ppm max of Class 3 residual solvents
  • 1 ppm max. of Class 1 Solvents, 10 ppm max. of Class 2 solvents, and 50 ppm max of Class 3 residual solvents
  • Processed for low water content to facilitate the extraction of organic volatile impurities, or VOC
TRUSTED_SUSTAINABILITY
Absorbance 0.100 AU Maximum (325nm), 0.030 AU Maximum (350nm), 0.010 AU Maximum (400nm)
CAS 872-50-4
Color Colorless to Yellow
Quantity 1 L
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Molecular Formula C5H9NO
Water 0.05% Maximum
Synonym 1-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidone, 1-methyl-2-pyrrolidone, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methylpyrrolidinone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl
InChI Key SECXISVLQFMRJM-UHFFFAOYSA-N
SMILES CN1CCCC1=O
IUPAC Name 1-methylpyrrolidin-2-one
Molecular Weight (g/mol) 99.133
PubChem CID 13387
ChEBI CHEBI:7307
Formula Weight 99.13
Percent Purity ≥99.9%
Grade GC Headspace
Chemical Name or Material NMP, N-Methylpyrrolidone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-methyl-2-pyrrolidone
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H319-Causes serious eye irritation.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contains : 1-methyl-2-pyrrolidone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'