missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Nonwoven Cleanroom Wipes

Fisherbrand™ Nonwoven Cleanroom Wipes are tough, abrasion resistant, absorbent, and virtually free of particle generation.

28.50 € - 29.05 €

Specifications

Certifications/Compliance ISO Class 5-6 (100-1000) Cleanroom Wipe, USP 797 and USP 800 wipe requirements
Color White
For Use With (Application) Laboratory and cleanroom
Material Polyester/Cellulose Blend
Product Type Cleanroom Wipes
View More Specs

Products 2
Product Code Brand Dimensions (L x W) Packaging Quantity Price Quantity & Availability  
Product Code Brand Dimensions (L x W) Packaging Quantity Price Quantity & Availability  
16398307 View Documents Fisherbrand™
9 x 9 in. (22.86 x 22.86 cm) 300 Wipes/Bag, 12 Bags/Case 28.50 €
Pack of 300 View alternative products
Save Up to 
»
In stock, ready to ship: 4
Add to basket
 
16388307 View Documents Fisherbrand™
12 x 12 in. (30.48 x 30.48 cm) 150 Wipes/Bag, 14 Bags/Case 29.05 €
Pack of 150 View alternative products
Save Up to 
»
In stock, ready to ship: 20
Add to basket
 
Description

Description

  • Made from a hydroentangled non-woven material composed of 45% polyester and 55% cellulose
  • Manufactured in a cleanroom environment and cleanroom packaged
  • Manufactured from poly-cellulose material providing a tough, absorbent and abrasion resistant product
  • Production process guarantees extremely low particle generation and chemical extractables
  • Compatible with virtually any solvent or solution for cleaning and wiping
  • Double bag packaging
  • Provide lot-to-lot traceability
Specifications

Specifications

ISO Class 5-6 (100-1000) Cleanroom Wipe, USP 797 and USP 800 wipe requirements
Laboratory and cleanroom
Cleanroom Wipes
White
Polyester/Cellulose Blend
Non-sterile