missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Norfloxacin, 98%, Analytical standard, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Type | Norfloxacin |
Physical Form | Crystalline Powder |
CAS | 70458-96-7 |
Melting Point | 218°C to 226°C |
Ignition Residue | 0.2 % max. |
Infrared Spectrum | Authentic |
Assay Percent Range | 98% |
Molecular Formula | C16H18FN3O3 |
Synonym | 1-Ethyl-6-fluoro-1, 4-dihydro-4-oxo-7-piperazino-3-quinolinecarboxylic acid |
InChI Key | OGJPXUAPXNRGGI-UHFFFAOYSA-N |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'