Learn More
Micronova™ NovaClean™ Lab & Glass Cleaner
This ready to use, low-ionic detergent can be applied directly to a variety of cleanroom work surfaces including glass, stainless steel, epoxy acrylic, and vinyl curtains. Available in an easy to use, 1 quart spray bottle and a 1 gallon bottle.
Brand: Micronova™ NC2-Q
310.08 EUR valid until 2024-04-30
Use promo code "21673" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Detergent for workstations and glass
The Micronova NovaClean™ Lab & Glass Cleaner is a low-ionic detergent, designed for use on cleanroom work-stations, laminar flow benches, acrylic and painted surfaces.
- Ready to use, cleanroom surfactant
- Filtered to 0.1 microns
- Low mobile ions
- Low non-volatile residue levels
- Can be applied directly to a range of surfaces
- Works well with glass, stainless steel, epoxy, acrylic, and vinyl curtains
- Ideal for conductive flooring
- Available in a 1 quart spray bottle and a 1 gallon refill bottle
- Biodegradable
- Free-rinsing
Specifications
0.1 μ Filtered Low Ionic Surfactant | |
Clear | |
Store at ambient temperature. Avoid direct sunlight. | |
1 qt. |
Non-sterile | |
Not Autoclavable | |
Mops and Buckets or Wipes and Sponge |