missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Bolt™ Transfer Buffer (20X)
Optimized for the transfer of proteins from Bolt Bis-Tris Plus gels to membranes for western blotting
43.20 €
Specifications
Content And Storage | Store at 4 to 25°C. |
---|---|
Buffer | Transfer Buffers |
Concentration | 20 X |
Product Type | Western Blot Transfer Buffer |
Form | Liquid |
Description
- When combined with 10% methanol, Bolt Transfer Buffer can be used with Bolt Mini Blot Module for wet transfer
- Can also be used with Bolt Antioxidant to enhance transfer of reduced proteins to membranes
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis, Western Blotting
Specifications
Store at 4 to 25°C. | |
20 X | |
Liquid | |
Bolt™ |
Transfer Buffers | |
Western Blot Transfer Buffer | |
Bolt™ Bis-Tris Plus Gels |
For Research Use Only. Not for use in diagnostic procedures.