missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ iBlot™ 2 Transfer Stacks, nitrocellulose, regular size

Used to transfer proteins with the iBlot 2 Gel Transfer Device

Brand:  Invitrogen™ IB23001

201.26 EUR valid until 2024-04-30
Use promo code "21741" to get your promotional price.



Additional Details : Weight : 1.00000kg


 View more versions of this product

Product Code. 15296526

  • 220.00 € / Pack of 10
In Stock
to see stock.
Add to basket

Description

Description

The iBlot 2 Transfer Stacks are consumable stacks with integrated nitrocellulose transfer membranes for dry blotting of proteins. Each stack contains a copper coated electrode and appropriate cathode and anode buffers in the gel matrix to allow fast, reliable transfer of proteins.

  • Increased shelf life
  • Reduced and compact packaging
  • Complete compatibility with iBlot 2 Gel Transfer Device
  • The nitrocellulose membrane (0.2μm pore size) in this stack is composed of 100% pure nitrocellulose to provide high-quality transfer
  • Membrane is compatible with commonly used detection methods such as staining, immunodetection, fluorescence, or radiolabeling
  • Protein binding capacity is 209μg/cm2

Transfer Stack Sizes

  • The regular size (13 x 8.3cm) of this stack is used for all Novex midi gels (or other midi gel types), two Novex mini gels (or other mini gel types), and E-PAGE™ 48 or 96 gels
  • The mini size (8 x 8cm) is used for all Novex mini gels (or other mini gel types)

Proteins, Expression, Isolation and Analysis, Western Blotting

Specifications

Specifications

13.5 cm
Transfer Stack
0.2 μm
Regular, 13.5 cm x 8 cm
10 Blots
8 cm
Nitrocellulose
Store at room temperature.
iBlot
Less waste, sustainable packaging, sustainable disposal
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • X10 iBlot® 2 Transfer Stacks, nitrocellulose

Signal Word
  • Warning

Hazard Category
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H373-May cause damage to organs.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P314-Get medical advice/attention if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Ethylene glycol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.