Learn More
Invitrogen™ Novex™ P-Selectin (soluble) Human ELISA Kit
Quantifies levels of soluble human P-selectin in cell culture supernatant, human serum, plasma, amniotic fluid, or other body fluids
Brand: Invitrogen™ KHS2021
Additional Details : Weight : 0.47000kg
Includes: Pre-coated plate, standard, control, antibody-HRP, diluents, wash buffer, chromogen, stop solution, color dyes, and plate covers.
Description
P-selectin acts as a receptor that supports binding of leukocytes.
Cell Adhesion, Cell Analysis, Cell Viability, Proliferation & Function, Immunoassays, Ready-To-Use Immunoassay
Order Info
Shipping Condition: Room Temperature
Specifications
NM_003005.3, NP_002996.2 | |
Amniotic Fluid, Plasma, Serum, Supernatant | |
Microplate Reader | |
2.25 hr. | |
P-Selectin | |
selectin P (granule membrane protein 140kDa, antigen CD62) |
<0.5ng/mL | |
Colorimetric Detection | |
6403 | |
Antibody-HRP, Chromogen, Color dyes, Control, Diluents, Plate covers, Pre-coated plate, Standard, Stop solution, Wash buffer | |
96 Assays |
For Research Use Only. Not for use in diagnostic procedures.