Learn More
Macherey-Nagel Bioanalysis™ NucleoSpin™ 96 Tissue, 96-Well Kit
Allround kit for purification of DNA from a broad range of samples in proven 96‐well plate format.
Brand: Macherey-Nagel Bioanalysis™ 740741.2
Code : LE
Additional Details : Weight : 0.39300kg Transport : UN number : 1993 Chem class : 3 Pack group : III
Description
NucleoSpin™ 96 Tissue is designed for the rapid simultaneous preparation of highly pure genomic DNA from animal and human tissue, eg. mouse and rat tails.
From a 1.2 cm mouse tail tip section (age of mice 4 to 6 weeks), up to 30 to 40 μg of pure genomic DNA can be prepared (typical yields 15 to 25 μg). The obtained DNA can be used directly for PCR, Southern blotting or any kind of enzymatic reaction.NucleoSpin™ Tissue binding plates can be used under vacuum or centrifugation in a centrifuge with deep buckets allowing processing of NucleoSpin™ 96 well plates stacked on 96 well blocks. Up to 192 samples (two plates) can be processed in less than one hour (after lysis). With the NucleoSpin™ 96 Tissue method, lysis is achieved by incubation of the samples in a solution containing detergents and proteinase K at 56°C. Appropriate conditions for binding of DNA to the silica membranes of the NucleoSpin™ Tissue binding plate are created by addition of large amounts of chaotropic ions plus ethanol to the lysate. The binding process is reversible and specific to nucleic acids. Contaminants are removed by washing with two different buffers. Pure genomic DNA is finally eluted under low ionic strength conditions in a slightly alkaline elution buffer.Kit components: NucleoSpin™ Tissue binding plates, buffers, proteinase K, wash plates, square-well blocks, round-well blocks, racks with tube strips, cap strips, self-adhesive foil.
- Silica membrane technology
- Format: 96 well plates
- Preparation time: 1 hr (after lysis)
- Sample material: ≤ 20 mg tissue, 106 cultured or bacteria
- Yield: 15 to 25 μg
- Elution volume: 100 to 200 μL
- Fragment size: 300 bp to 30 kb
- No use of organic solvents
Specifications
18°C to 25°C | |
96-well Plate | |
Isolation of DNA | |
192 Preps |
Safety and Handling
- Danger
Hazard Category
- Flammable liquid Category 3
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H336-May cause drowsiness or dizziness.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
- MIXTURE LIST-Contains : guanidine hydrochloride, 2-propanol