missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ gDNA Clean-up XS, Micro kit for DNA clean up and concentration

Product Code. p-8013746
Change view
Click to view available options
No. of Reactions:
10
50
Unit Size:
10 reactions
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
12733411 10 10 reactions
12753411 50 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12733411 Supplier Macherey-Nagel™ Supplier No. 740904.10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Effective post clean up and concentration of genomic DNA.

Specifications

Detection Method Silica-membrane Technology
Format Mini Spin Column - XS Design
For Use With (Application) Small DNA Quantities Extracted
Includes 2mL Collection Tubes, Buffers
No. of Reactions 10
Product Type NucleoSpin™ gDNA Clean-up XS Column
Quantity 1 Each
Sample Volume <400 μL solution containing <2 μg DNA
Test Time 20min.
Yield 60 to 70%
compliance-icons
Product Identifier
  • NucleoSpin gDNA Clean-up XS (10)
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.
Supplemental information
  • MIXTURE LIST-Contain: guanidinium thiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.