Learn More
Thermo Scientific™ Nunc™ 384-Well, Non-Treated, Flat-Bottom Microplate
Color:Clear; Sterility:Sterile; Lid:Without Lid
Brand: Thermo Scientific™ 265203
Code : 76
Additional Details : Weight : 7.53000kg
Description
Thermo Scientific™ Nunc 384-Well Clear Polystyrene Plates with Cell Culture Treated and Nontreated Surfaces are optimized for robotics, plate readers and liquid handling systems. 384-well microplates allow sample and reagent volume reduction providing cost savings and allowing more samples to be screened. Our plates are high quality for consistent and reliable results.
Nunc 384-well polystyrene plates have rounded square-well geometry, eliminating wicking.- Clear Polystyrene has excellent optical properties making it ideal for optical measurements
- Untreated Polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
- Untreated surfaces are ideal for adding coatings in your lab
- Cell Culture Treated: The Nunclon Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
- Customized bar coding is available
- Flexible working volumes; 10-100μL
Specifications
Custom Barcoding Available | |
Without Lid | |
Non-Treated; no lid; Sterile; 25/100 | |
25/Pk., 100/Cs. | |
Clear | |
Polystyrene | |
384 | |
Sterile | |
Flat Bottom (F-well) | |
25 |
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) | |
0.084 cm2/well | |
16 x 24 | |
Nunc | |
Microplate | |
120 μL | |
10 to 100 μL | |
Non-treated | |
100 |