missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ Cell Culture Inserts in Carrier Plate Systems

Product Code. 11842700
Click to view available options
Pore Size:
0.4 μm
3 μm
8 μm
No Inserts
No. of Wells:
12
24
Unit Size:
Pack of 4
Pack of 48
Pack of 96
This item is not returnable. View return policy

Product Code. 11842700

Brand: Thermo Scientific™ 141006

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Use up to 24 inserts at a time with Thermo Scientific™ Nunc™ Carrier Plate for Cell Culture Inserts, for individual compartment cultures or cultures with shared media.

The Thermo Scientific™ Nunc™ Carrier Plate system is designed to enable three different hanging height adjustments for the Cell Culture Inserts to accommodate liquid-air-interface culture and various cell feeding protocols. Individually packed, the multi-well Carrier Plate system comes with or without the Cell Culture Inserts.

Features:
  • Meets ANSI™ standards
  • Equipped with alphanumeric well identification
  • Polycarbonate membrane is available in three pore sizes for enhanced flexibility in your research.
  • Inserts are ideal for for 3-D imaging; translucent membrane becomes transparent when wet.
  • Carrier plates come in two formats that correspond with our 12- and 24-well Thermo Scientific™ Nunclon™ Delta-treated dishes.
  • Uniquely engineered carrier plate features a support structure that enables variable height.
  • Simply rotate the insert to engage one of the three height-adjusting tabs into the carrier plate supports.
TRUSTED_SUSTAINABILITY

Spezifikation

Inserts per Plate 24
Culture Area 0.47 cm2
Material Polycarbonate (Membrane)
Volume (Metric) Working 1 mL (Low Hanging Position), 1.5 mL (Medium Hanging Position), 2 mL (High Hanging Position)
Pore Density <0.85 x 105 pores/cm2
Pore Size 8 μm
Type Carrier Plate with Cell Culture Inserts
For Use With Cell Culture Insert
No. per Pack 1
No. per Case 4
Dish Type Carrier Plate Systems
Color Clear
Surface Treatment Cell culture-treated
Sterility Sterile
No. of Wells 24
Mehr anzeigen Weniger anzeigen

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt