Learn More
Cytiva (formerly Pall Lab) Nylaflo™ Nylon Membrane Disc Filters
A highly versatile laboratory membrane filter.
Brand:  Cytiva (formerly Pall Lab) 66604
Additional Details : Weight : 0.00100kg
 View more versions of this product
Product Code. 17104591
  • 374.00 € / Pack of 25
Estimated Shipment: 30-07-2024
to see stock.
Add to basket
Description
Description
  • Chemical compatibility with esters, bases, and alcohols naturally hydrophilic
  • HPLC certified
  • Provides assurance that the filter will not add artifacts to your analysis
  • For research use only
Specifications
Specifications
Nylon (Hydrophilic)
Non-sterile
127 μm
0.2 μm
25/Pk.
142 mm