missing translation for 'onlineSavingsMsg'
Learn More

Biodesign™ Nylon Matrix Filter CellMicroSieves™

Product Code. 12974257
Change view
Click to view available options
Pore Size:
5 μm
10 μm
15 μm
20 μm
25 μm
30 μm
35 μm
50 μm
70 μm
100 μm
200 μm
Quantity:
1 Ea.
1 Roll
10 Pk.
3 Pk.
Unit Size:
Each
Pack of 10
Pack of 3
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size Quantity unitSize
12974257 10 μm 3 Pk. Pack of 3
12684787 100 μm 3 Pk. Pack of 3
12914277 200 μm 10 Pk. Pack of 10
12964257 15 μm 3 Pk. Pack of 3
12904267 200 μm 1 Roll Each
12994247 200 μm 3 Pk. Pack of 3
12954257 20 μm 3 Pk. Pack of 3
12944257 25 μm 3 Pk. Pack of 3
12987487 30 μm 1 Ea. Each
12944267 35 μm 1 Roll Each
12924257 50 μm 3 Pk. Pack of 3
12904277 5 μm 1 Roll Each
12994257 5 μm 3 Pk. Pack of 3
12914257 70 μm 3 Pk. Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
This item is not returnable. View return policy
Product Code. 12974257 Supplier Biodesign™ Supplier No. N10S
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Allow the rapid, yet gentle, isolation of cells, organelles, or cellular debris.

Unlike traditional filters, CellMicroSieves™ are a precision woven matrix of pure nylon filaments that is available with pores ranging from 5 μm to 200 μm. CellMicroSieves™ do not wet and contains no dyes, detergents or extractables. Extremely low protein affinity means that cells and proteins will not bind to CellMicroSieves™. They have a wide variety of distinctive uses a specific sized porous screens in the laboratory. AccessoriesPolypropylene sieve holders (2 piece) holders with adjustable metal tightener or filtration units, type A borosilicate glass with Stainless Steel filter support and aluminum clamp or type B complete polysulfone filtration unit with polypropylene filter support are available. Both filtration units have a 300mL capacity are autoclavable and reusable. Use sheets and rolls with sieve holders and circles with filtration units to hold securely when processing samples.

  • A pure nylon product that is biologically inert, resistant to most acidic or basic conditions and many solvents
  • Autoclavable and can be repeatedly washed and reused
  • Rapid collection of tissue culture cells without centrifugation
  • Isolation of specific cell types from a dissociated tissue sample, allowing only a single type of cell to pass through
  • Separation of different sized cells from a mixture of cell types
  • Concentration and isolation of cells from a bioreactor
  • Prefilter to prevent clogging of final filter CellMicroSieves™ allow the rapid gentle isolation of cells, organelles or cellular debris.

Specifications

Quantity 3 Pk.
Pore Size 10 μm
Type Cell Micro Sieve
Material Nylon
For Use With (Application) For rapid collection of tissue culture cells without centrifugation
Autoclavable Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.