missing translation for 'onlineSavingsMsg'
Learn More

O-(N-Succinimidyl)-N,N,N',N'-tetramethyluronium tetrafluoroborate, 97%, Thermo Scientific Chemicals

Catalog Number 11975581
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
105832-38-0
C9H16BF4N3O3
301.05
MFCD00077875
YEBLHMRPZHNTEK-UHFFFAOYSA-N
tstu, o-n-succinimidyl-1,1,3,3-tetramethyluronium tetrafluoroborate, 2-2,5-dioxopyrrolidin-1-yl-1,1,3,3-tetramethylisouronium tetrafluoroborate, 2-succinimido-1,1,3,3-tetramethyluronium tetrafluoroborate, n,n,n',n'-tetramethyl-o-n-succinimidyl uronium tetrafluoroborate, o-n-succinimidyl-n,n,n',n'-tetramethyluronium tetrafluoroborate, o-n-succinimidyl-n,n,n',n'-tetramethyl-uronium tetrafluoroborate, dimethylamino 2,5-dioxopyrrolidin-1-yl oxy methylidene dimethylazanium tetrafluoroborate, o-n-succinimidyl-n,n,n',n'-tetramethyl uronium bf4, pubchem12781
9857522
[dimethylamino-(2,5-dioxopyrrolidin-1-yl)oxymethylidene]-dimethylazanium;tetrafluoroborate
F[B-](F)(F)F.CN(C)C(ON1C(=O)CCC1=O)=[N+](C)C
This item is not returnable. View return policy
105832-38-0
C9H16BF4N3O3
301.05
MFCD00077875
YEBLHMRPZHNTEK-UHFFFAOYSA-N
tstu, o-n-succinimidyl-1,1,3,3-tetramethyluronium tetrafluoroborate, 2-2,5-dioxopyrrolidin-1-yl-1,1,3,3-tetramethylisouronium tetrafluoroborate, 2-succinimido-1,1,3,3-tetramethyluronium tetrafluoroborate, n,n,n',n'-tetramethyl-o-n-succinimidyl uronium tetrafluoroborate, o-n-succinimidyl-n,n,n',n'-tetramethyluronium tetrafluoroborate, o-n-succinimidyl-n,n,n',n'-tetramethyl-uronium tetrafluoroborate, dimethylamino 2,5-dioxopyrrolidin-1-yl oxy methylidene dimethylazanium tetrafluoroborate, o-n-succinimidyl-n,n,n',n'-tetramethyl uronium bf4, pubchem12781
9857522
[dimethylamino-(2,5-dioxopyrrolidin-1-yl)oxymethylidene]-dimethylazanium;tetrafluoroborate
F[B-](F)(F)F.CN(C)C(ON1C(=O)CCC1=O)=[N+](C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 194.0°C to 200.0°C
Color White
Elemental Analysis 34.47 to 37.34% C, 13.40 to 14.52% N
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Formula Weight 301.05
Percent Purity 97%
Physical Form Crystalline Powder or Needles
Chemical Name or Material O-(N-Succinimidyl)-N,N,N',N'-tetramethyluronium tetrafluoroborate
Show More Show Less
compliance-icons
Product Identifier
  • O-(N-Succinimidyl)-N,N,N',N'-tetramethyluronium tetrafluoroborate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'