missing translation for 'onlineSavingsMsg'
Learn More

o-Phenylenediamine dihydrochloride, 99+%, Thermo Scientific Chemicals

Catalog Number p-183020
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
615-28-1
C6H8N2·2HCl
181.07
MFCD00012966
RIIWUGSYXOBDMC-UHFFFAOYSA-N
1,2-phenylenediamine dihydrochloride, o-phenylenediamine dihydrochloride, 1,2-benzenediamine, dihydrochloride, opd easy-tablets, opd tablets, benzene-1,2-diamine dihydrochloride, usaf ek-678, o-phenylenediamine.2hcl, 1,2-benzenediamine dihydrochloride, 1,2-diaminobenzene dihydrochloride
11990
benzene-1,2-diamine;dihydrochloride
C1=CC=C(C(=C1)N)N.Cl.Cl
This item is not returnable. View return policy
615-28-1
C6H8N2·2HCl
181.07
MFCD00012966
RIIWUGSYXOBDMC-UHFFFAOYSA-N
1,2-phenylenediamine dihydrochloride, o-phenylenediamine dihydrochloride, 1,2-benzenediamine, dihydrochloride, opd easy-tablets, opd tablets, benzene-1,2-diamine dihydrochloride, usaf ek-678, o-phenylenediamine.2hcl, 1,2-benzenediamine dihydrochloride, 1,2-diaminobenzene dihydrochloride
11990
benzene-1,2-diamine;dihydrochloride
C1=CC=C(C(=C1)N)N.Cl.Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Melting Point 258°C
Color Pink to Purple
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula C6H4(NH2)2·2HCl
Quantity 25 g
Beilstein 13, III, 28
Formula Weight 181.07
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material o-Phenylenediamine dihydrochloride, 99+%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'