Learn More
Cytiva O-ring
Use in aqueous solutions and nearly all organic solvents commonly used in liquid chromatography of macromolecules. Cytiva Life Sciences™ O-rings are made of nitrile rubber.
Brand:  Cytiva 19-0685-01
Code : GE
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 10201640
  • 94.00 € / Pack of 5
Estimated Shipment: 05-07-2024
to see stock.
Add to basket
Description
Description
  • Can be used in aqueous solutions
  • Nearly all organic solvents
  • The following few exceptions: chlorinated hydrocarbons, acetone and otherketones, aliphatic esters and phenol. Solutions containingmore than 10% NaOH, 10% HCl, other strong mineral acidsor 5% acetic acid should not be used
  • Made of Nitrile Rubber (NBR)
Specifications
Specifications
O-Ring
Nitrile Rubber
For use with Series XK and K 9
1Each
37.47 x 5.34mm