Learn More
DWK Life Sciences O-ring, for stopcock keys with groove
For securing the DURAN™ valve plug with groove in the DURAN™ single way stopcock.
Brand: DWK Life Sciences 286031408
Additional Details : Weight : 0.05000kg
Description
The O-ring in the groove of the DURAN™ tap plug provided for this purpose secures the tap plug in the disposable tap.
- Secures the valve plug from falling out of the valve
Specifications
24.0 NS | |
Sealing | |
100/Pk. |