missing translation for 'onlineSavingsMsg'
Learn More

o-Tolidine dihydrochloride, 99%, ACROS Organics™

Brand:  Acros Organics 421135000

Code : 29

Additional Details : CAS Number : 612-82-8 Weight : 0.55000kg Transport : UN number : 3077 Chem class : 9 Pack group : III

Packaging Glass Bottle
Quantity 500g

 View more versions of this product

Product Code. 10015660

  • 399.00 € / 500g

Estimated Shipment
11-02-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

612-82-8
285.2
3,3'-dimethylbenzidine dihydrochloride, o-tolidine dihydrochloride, 3,3'-dimethyl-1,1'-biphenyl-4,4'-diamine dihydrochloride, unii-5msk350kd8, dsstox_cid_511, dsstox_rid_75633, dsstox_gsid_20511, tolidine dihydrochloride, 1,1'-biphenyl-4,4'-diamine, 3,3'-dimethyl-, dihydrochloride, 3,3'-dimethylbenzidine.2hcl
4-(4-amino-3-methylphenyl)-2-methylaniline;dihydrochloride
C14H16N2·2HCl
LUKPNZHXJRJBAN-UHFFFAOYSA-N
108938
CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)C)N.Cl.Cl

Specifications

o-Tolidine dihydrochloride
Glass Bottle
Beige to Beige-Gray
500g
98.5% min. (on dry substance) (Argentometry)
15,9675
LUKPNZHXJRJBAN-UHFFFAOYSA-N
4-(4-amino-3-methylphenyl)-2-methylaniline;dihydrochloride
108938
Crystalline Powder
Authentic
5% Max. (K.F.)
130.0°C
612-82-8
C14H16N2·2HCl
3,3'-dimethylbenzidine dihydrochloride, o-tolidine dihydrochloride, 3,3'-dimethyl-1,1'-biphenyl-4,4'-diamine dihydrochloride, unii-5msk350kd8, dsstox_cid_511, dsstox_rid_75633, dsstox_gsid_20511, tolidine dihydrochloride, 1,1'-biphenyl-4,4'-diamine, 3,3'-dimethyl-, dihydrochloride, 3,3'-dimethylbenzidine.2hcl
CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)C)N.Cl.Cl
285.2
285.2
99%
Videos
SDS Documents