Learn More
Macherey-Nagel™ Octadecyl Phase Polygosil™ C18
Brand:  Macherey-Nagel™ 711330.100
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10641292
  • 1755.00 € / 100g
Estimated Shipment: 18-06-2024
to see stock.
Add to basket
Specifications
5 μm
Liquid Chromatography
Polygosil C18 Phases
60 Å
100 g