missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Octagonal Shape PTFE Magnetic Stirring Bars

PTFE construction. Angular surface creates high turbulence even at low speeds.

Brand:  BRAND™ 137607

Code : A0

Additional Details : Weight : 0.22300kg

 View more versions of this product

Product Code. 12030932

  • 45.30 € / Pack of 10
In Stock
to see stock.
Add to basket

Description

Description

  • Octagonal shape
  • For vessels with slightly uneven bottoms.
  • Pivot ring reduces the contact surface which enables the stirrer to adopt the optimum position.
Specifications

Specifications

8 mm
PTFE
13 mm
10/Pk.
For vessels with slightly uneven bottoms
Octagon
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers