missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Octopamine hydrochloride, 99%, Thermo Scientific™

alpha-adrenoceptor agonist; invertebrate neurotransmitter and neuromodulator

26.20 € - 116.00 €


Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15423899
SDS View Documents
Thermo Scientific Alfa Aesar
J61281.03
26.20 €
1g
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
15433899
SDS View Documents
Thermo Scientific Alfa Aesar
J61281.06
69.50 €
5g
View alternative products
Save Up to 
»
Estimated Shipment: 16-09-2026
Add to basket
 
 Restricted Item
15443899
SDS View Documents
Thermo Scientific Alfa Aesar
J61281.09
10g
116.00 €
10g
View alternative products
Save Up to 
»
Estimated Shipment: 10-09-2026
Add to basket
 
 Restricted Item
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Octopamine HCL has the ability to halt catabolism of proteinaceous tissues. It is very important while losing weight to maximize adipose loss while minimizing the loss of muscle and other lean body tissue. Octopamine Hcl (norepinephrine) is best known as an effective ephedrine alternative, claiming to boost energy while reduce weight in bodybuilding industry for weight loss purpose. In addition, many users believe octopamine hcl is a good nootropic compound that makes our brain easy to focus, enhances motivation as a wakefulness promoter.

Solubility
It is soluble in water.

Notes
Handle under argon. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Videos
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.