Learn More
OHAUS™ Bench Scales Computer Data Transfer Cables
Connects Link Champ General-Purpose Bench Scales to IBM-PC computers via standard RS-232-C.
Brand: OHAUS™ 12106964
Code : A0
Additional Details : Weight : 0.15900kg
Description
Permits data collection via computer.
Specifications
Models B10S05, B25S05, B50S05, B100S05, B150S05 | |
RS-232 cable, PC 9 pin | |
Computer data transfer |