missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius™ OIML Class E2 Proof Line Knob Weight with DAkkS Certificate
Polished non-magnetizable calibration weights. Sartorius™ OIML Class E2 Proof Line Knob Weight with DAkkS Certificate is constructed of special steel and supplied in a plastic case.
164.00 € - 1560.00 €
Specifications
Product Type | Proof Line Knob Weight |
---|---|
Certifications/Compliance | DAkkS calibration certificate |
Classification | OIML Class E2 |
Material | Special Stainless Steel |
Density | 7,950kg/m3 |
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
11303314
|
Sartorius™
YCW312-AC-02 |
1 g |
164.00 €
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
11313314
|
Sartorius™
YCW322-AC-02 |
2 g |
172.00 €
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
11323314
|
Sartorius™
YCW352-AC-02 |
5 g |
177.00 €
Each |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
11333314
|
Sartorius™
YCW412-AC-02 |
10 g |
183.00 €
Each |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
11343314
|
Sartorius™
YCW422-AC-02 |
20 g |
195.00 €
Each |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
11353314
|
Sartorius™
YCW452-AC-02 |
50 g |
211.00 €
Each |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
11363314
|
Sartorius™
YCW512-AC-02 |
100 g |
236.00 €
Each |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
11373314
|
Sartorius™
YCW522-AC-02 |
200 g |
273.00 €
Each |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
11383314
|
Sartorius™
YCW552-AC-02 |
500 g |
383.00 €
Each |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
11393314
|
Sartorius™
YCW612-AC-02 |
1 kg |
535.00 €
Each |
In Stock
Log in to see stock. |
|||||
11303324
|
Sartorius™
YCW622-AC-02 |
2 kg |
775.00 €
Each |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
11313324
|
Sartorius™
YCW652-AC-02 |
5 kg |
1100.00 €
Each |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
11357494
|
Sartorius™
YCW712-AC-02 |
10 kg |
1560.00 €
Each |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
Description
- Non-magnetizable
- Supplied in a high-quality plastic case (unless otherwise specified) suitable for use in cleanrooms
- Special Stainless steel construction with a density of 7,950 kg/m3
- Complies with OIML R111-1 International Recommentation
- DAkkS Certified
Specifications
Proof Line Knob Weight | |
OIML Class E2 | |
7,950kg/m3 |
DAkkS calibration certificate | |
Special Stainless Steel |