missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius™ OIML Class E2 Proof Line Knob Weight without Certificate
Polished non-magnetizable calibration weights. Sartorius™ OIML Class E2 Knob Weight without Certificate is constructed of special steel and supplied in a plastic case.
80.60 € - 1480.00 €
Specifications
Product Type | Proof Line Knob Weight |
---|---|
Classification | OIML Class E2 |
Material | Special Stainless Steel |
Density | 7,950kg/m3 |
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
11363284
|
Sartorius™
YCW312-AC-00 |
1 g |
80.60 €
Each |
Estimated Shipment: 02-05-2024
Log in to see stock. |
|||||
11373284
|
Sartorius™
YCW322-AC-00 |
2 g |
88.70 €
Each |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
11383284
|
Sartorius™
YCW352-AC-00 |
5 g |
93.80 €
Each |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
11393284
|
Sartorius™
YCW412-AC-00 |
10 g |
98.90 €
Each |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
11303294
|
Sartorius™
YCW422-AC-00 |
20 g |
111.00 €
Each |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
11313294
|
Sartorius™
YCW452-AC-00 |
50 g |
128.00 €
Each |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
11323294
|
Sartorius™
YCW512-AC-00 |
100 g |
152.00 €
Each |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
11333294
|
Sartorius™
YCW522-AC-00 |
200 g |
190.00 €
Each |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
11343294
|
Sartorius™
YCW552-AC-00 |
500 g |
299.00 €
Each |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
11353294
|
Sartorius™
YCW612-AC-00 |
1 kg |
448.00 €
Each |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
11363294
|
Sartorius™
YCW622-AC-00 |
2 kg |
690.00 €
Each |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
11373294
|
Sartorius™
YCW652-AC-00 |
5 kg |
1010.00 €
Each |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
11343434
|
Sartorius™
YCW712-AC-00 |
10 kg |
1480.00 €
Each |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
Description
- Non-magnetizable
- Supplied in a high-quality plastic case (unless otherwise specified) suitable for use in cleanrooms
- Special Stainless steel construction with a density of 7,950 kg/m3
- Complies with OIML R111-1 International Recommentation
Specifications
Proof Line Knob Weight | |
OIML Class E2 | |
Special Stainless Steel | |
7,950kg/m3 |