missing translation for 'onlineSavingsMsg'
Learn More

Oleic acid, 99%, Thermo Scientific Chemicals

Catalog Number 11398307
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
112-80-1
C18H34O2
282.47
MFCD00064242
ZQPPMHVWECSIRJ-MDZDMXLPSA-N
oleic acid, cis-9-octadecenoic acid, cis-oleic acid, elaidoic acid, oleate, glycon wo, wecoline oo, pamolyn 100, glycon ro, z-octadec-9-enoic acid
445639
CHEBI:16196
(Z)-octadec-9-enoic acid
CCCCCCCC\C=C\CCCCCCCC(O)=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
112-80-1
C18H34O2
282.47
MFCD00064242
ZQPPMHVWECSIRJ-MDZDMXLPSA-N
oleic acid, cis-9-octadecenoic acid, cis-oleic acid, elaidoic acid, oleate, glycon wo, wecoline oo, pamolyn 100, glycon ro, z-octadec-9-enoic acid
445639
CHEBI:16196
(Z)-octadec-9-enoic acid
CCCCCCCC\C=C\CCCCCCCC(O)=O

Oleic acid is used as an emulsifying agent for facial creams and lotions, as an accelerator in the rubber industry, as an emollient, as a solubilizing agent in aerosol products and as a soldering flux in stained glass work for joining lead came. It is used in the preparation of oleates and lotions. It acts as intermediate for drilling and fracking fluids in the oil and gas industry. It finds application as an excipient in pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 13°C to 14°C
Color Colorless to Yellow
Density 0.887 g/mL
Boiling Point 286°C (100 mmHg)
Refractive Index 1.461
Quantity 5 g
Beilstein 1726542
Sensitivity Air Sensitive
Merck Index 146828
Solubility Information Miscible with ethanol,ether,acetone,chloroform,dimethyl formamide and dimethyl sulfoxide.
Formula Weight 282.47
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Oleic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'